Isorhapontigenin 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-5-(4-hydroxy-3-methoxystyryl)benzene-1,3-diol, 3,4',5-Trihydroxy-3'-methoxy-trans-stilbene
A531729-10MG
A531729-1MG
A531729-5MG
77034-758EA
63.66
USD
77034-758
77034-760
77034-762
Isorhapontigenin 95%
Isorhapontigenin
Caution: for Research Use Only
Formula:
C₁₅H₁₄O₄ MW: 258.27 g/mol |
MDL Number:
MFCD12407151 CAS Number: 32507-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...