N,O-Bis(trimethylsilyl)acetamide 95%

Supplier: Ambeed
Danger

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

A470462-25G A470462-10G A470462-100G
77034-276EA 6.37 USD
77034-276 77551-434 77602-046
N,O-Bis(trimethylsilyl)acetamide 95%
N,O-Bis(trimethylsilyl)acetamide

Caution: for Research Use Only

Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR