Coenzyme Q10 97%

Supplier: Ambeed
Warning

Synonyms: (all-E)-2-(3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, Ubiquinone 10, Ubidecarenone

A422056-100G A422056-100MG A422056-1G A422056-250MG A422056-25G A422056-5G
77004-118EA 248.26 USD
77004-118 77004-120 77004-122 77004-124 77004-126 77004-128
Coenzyme Q10 97%
Coenzyme Q10

Caution: for Research Use Only

Formula: C₅₉H₉₀O₄
MW: 863.37 g/mol
MDL Number: MFCD00042919
CAS Number: 303-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR