Xanthotoxin 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-Methoxy-7H-furo[3,2-g]chromen-7-one, 8-Methoxypsoralen, Methoxsalen
A372661-10G
A372661-10MG
A372661-1G
A372661-1MG
A372661-25G
A372661-50MG
A372661-5G
A372661-100G
77003-512EA
48
USD
77003-512
77003-514
77003-516
77003-518
77003-520
77003-522
77003-524
77672-864
Xanthotoxin 98+%
Xanthotoxin
Caution: for Research Use Only
Formula:
C₁₂H₈O₄ MW: 216.19 g/mol |
MDL Number:
MFCD00005009 CAS Number: 298-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...