L-4-Cyanophenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Cyano-L-phenylalanine, H-Phe(4-CN)-OH, (2S)-2-Amino-3-(4-cyanophenyl)propanoic acid

A348964-1G A348964-250MG A348964-5G
77003-236EA 23.34 USD
77003-236 77003-238 77003-240
L-4-Cyanophenylalanine 98%
L-4-Cyanophenylalanine

Caution: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
MDL Number: MFCD00270363
CAS Number: 167479-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR