N-(2-Bromobenzyloxycarbonyloxy)succinimide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromobenzyl Succinimidyl Carbonate, Carbonic Acid 2-Bromobenzyl Succinimidyl Ester, Z(2-Br)-OSu
A324435-100G
A324435-25G
77002-910EA
82
USD
77002-910
77002-912
77002-914
N-(2-Bromobenzyloxycarbonyloxy)succinimide 98%
N-(2-Bromobenzyloxycarbonyloxy)succinimide
Caution: for Research Use Only
Formula:
C₁₂H₁₀BrNO₅ MW: 328.12 g/mol |
MDL Number:
MFCD00082848 CAS Number: 128611-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...