Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
A312950-1G
A312950-250MG
A312950-5G
77015-174EA
19
USD
77015-174
77015-176
77015-178
Ethyl-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Caution: for Research Use Only
Formula:
C₁₁H₈ClF₃N₂O₂ MW: 292.65 g/mol |
MDL Number:
MFCD01833013 CAS Number: 353258-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...