5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine 97%

Supplier: Ambeed
Warning

A307899-100G A307899-1G A307899-250MG A307899-25G A307899-5G
77002-630EA 351 USD
77002-630 77002-632 77002-634 77002-636 77002-638
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine 97%
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine

Caution: for Research Use Only

Formula: C₄H₇N₃OS
MDL Number: MFCD01203002
CAS Number: 15884-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR