PIPES sesquisodium salt 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid sesquisodium salt, Sesquisodium piperazine-1,4-diethanesulphonate, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulfonic) acid sesquisodium salt, Sodium 1,1'-(piperazine-1,4-diyl)diethanesulphonate 1-(4-(1-sulphoethyl)piperazin-1-yl)ethanesulphonate(3:2), Sesquisodium piperazine-1,4-diethanesulfonate, PIPES-Na1,5, Piperazine-N,N'-bis-2-ethanesulphonic acid sesquisodium salt, Piperazine-1,4-bis(2-ethanesulphonic acid) sesquisodium salt, Sodium 1,1'-(piperazine-1,4-diyl)diethanesulfonate 1-(4-(1-sulfoethyl)piperazin-1-yl)ethanesulfonate(3:2), Piperazine-1,4-bis(2-ethanesulfonic acid) sesquisodium salt, Piperazine-N,N'-bis-2-ethanesulfonic acid sesquisodium salt

A307624-10G A307624-25G
77032-550EA 8.49 USD
77032-550 77032-552
PIPES sesquisodium salt 98%
PIPES sesquisodium salt

Caution: for Research Use Only

Formula: C₁₆H₃₃N₄Na₃O₁₂S₄
MW: 670.69 g/mol
MDL Number: MFCD00066393
CAS Number: 100037-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR