Bicyclo[3.2.0]hept-2-en-6-one 95%

Supplier: Ambeed
Danger

Synonyms: (±)-cis-Bicyclo[3.2.0]hept-2-en-6-one

A305559-10G A305559-1G A305559-25G A305559-5G
77002-572EA 84 USD
77002-572 77002-574 77002-576 77002-578
Bicyclo[3.2.0]hept-2-en-6-one 95%
Bicyclo[3.2.0]hept-2-en-6-one

Caution: for Research Use Only

Formula: C₇H₈O
MW: 108.14 g/mol
MDL Number: MFCD00066201
CAS Number: 13173-09-6
UN: 1224
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR