(S)-tert-Butyl-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl-(S)-(3-amino-2-oxo-2,3,4,5-tetrahydrobenzo[b]azepin-1-yl) acetate, Benazepril Related Compound F, (S)-tert-Butyl 2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate

A280526-100G A280526-1G A280526-5G A280526-10G A280526-25G
77002-142EA 213 USD
77002-142 77002-144 77002-146 77672-442 77672-444
(S)-tert-Butyl-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate 97%
(S)-tert-Butyl-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate

Caution: for Research Use Only

Formula: C₁₆H₂₂N₂O₃
MDL Number: MFCD03426719
CAS Number: 109010-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR