2,4-Difluoro-3-cyanophenylboronic acid 95%

Supplier: Ambeed
Warning

A273538-1G A273538-250MG A273538-5G A273538-10G
77031-976EA 21 USD
77031-976 77031-978 77593-688 77601-196
2,4-Difluoro-3-cyanophenylboronic acid 95%
2,4-Difluoro-3-cyanophenylboronic acid

Caution: for Research Use Only

Formula: C₇H₄BF₂NO₂
MDL Number: MFCD12026699
CAS Number: 871940-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR