3-Amino-2,6-dimethoxypyridine 98%
Supplier: Ambeed
|
Warning
|
A261272-1G
A261272-250MG
A261272-5G
A261272-25G
77000-940EA
91
USD
77000-940
77000-942
77601-104
77881-707
3-Amino-2,6-dimethoxypyridine 98%
3-Amino-2,6-dimethoxypyridine
Caution: for Research Use Only
Formula:
C7H10N2O2 |
MDL Number:
MFCD00832865 CAS Number: 28020-37-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...