Glycitein 98%

Supplier: Ambeed
Warning

Synonyms: 7-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one, 7,4'-Dihydroxy-6-methoxyisoflavone

A231798-1MG A231798-5MG
77031-068EA 14 USD
77031-068 77031-070
Glycitein 98%
Glycitein

Caution: for Research Use Only

Formula: C₁₆H₁₂O₅
MW: 284.27 g/mol
MDL Number: MFCD00016679
CAS Number: 40957-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR