4-Fluoro-3-methylbenzamide 95%

Supplier: Ambeed
Warning

A199247-1G A199247-5G
76999-262EA 36 USD
76999-262 76999-264
4-Fluoro-3-methylbenzamide 95%
4-Fluoro-3-methylbenzamide

Caution: for Research Use Only

Formula: C₈H₈FNO
MW: 153.16 g/mol
MDL Number: MFCD01631489
CAS Number: 261945-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR