GSK1120212 98%

Supplier: Ambeed
Danger

Synonyms: N-(3-(3-Cyclopropyl-5-((2-fluoro-4-iodophenyl)amino)-6,8-dimethyl-2,4,7-trioxo-3,4,6,7-tetrahydropyrido[4,3-d]pyrimidin-1(2H)-yl)phenyl)acetamide, Trametinib, JTP-74057

A188937-100MG A188937-1G A188937-250MG
76986-032EA 95 USD
76986-032 76986-034 77567-762 77890-168
GSK1120212 98%
GSK1120212

Caution: For research use only.

Formula: C₂₆H₂₃FIN₅O₄
MDL Number: MFCD17215075
CAS Number: 871700-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR