Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Lys(Boc)-OH.DCHA, Boc-Lys(Boc)-OH·DCHA, Nα,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, Nɑ,N-ε-Di-Boc-L-lysine Dicyclohexylammonium Salt
A180992-10G
A180992-25G
A180992-5G
A180992-100G
A180992-500G
76998-672EA
12
USD
76998-672
76998-674
76998-676
77671-550
77671-552
Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt 96%
Nalpha,N-epsilon-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt
Caution: for Research Use Only
Formula:
C₂₈H₅₃N₃O₆ MW: 527.75 g/mol |
MDL Number:
MFCD00038892 CAS Number: 15098-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...