m-Cresol purple indicator
Supplier: Ambeed
|
Warning
|
Synonyms:
m-Cresolsulfonphthalein, 2',2''-Dimethylphenolsulphonphthalein, m-Cresolsulphonphthalein, MCP, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 2',2''-Dimethylphenolsulfonphthalein
A172369-100G
A172369-25G
A172369-5G
76985-500EA
97
USD
76985-500
76985-502
76985-504
m-Cresol purple indicator
m-Cresol purple
Caution: for Research Use Only
Formula:
C₂₁H₁₈O₅S MW: 382.43 g/mol |
MDL Number:
MFCD00005871 CAS Number: 2303-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...