N6-Benzoyladenine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Benzamidopurine, N-(7H-Purin-6-yl)benzamide
A170278-100G
A170278-10G
A170278-25G
A170278-500G
A170278-5G
77028-996EA
56
USD
77028-996
77028-998
77029-000
77029-002
77029-004
N6-Benzoyladenine 98%
N6-Benzoyladenine
Caution: for Research Use Only
Formula:
C₁₂H₉N₅O MW: 239.24 g/mol |
MDL Number:
MFCD00037927 CAS Number: 4005-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...