PQ401 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea, N-(5-Chloro-2-methoxyphenyl)-N’-(2-methyl-4-quinolinyl)urea, IGF-IR Inhibitor

A165138-10MG A165138-1MG A165138-50MG A165138-5MG
76985-212EA 55 USD
76985-212 76985-214 76985-216 76985-218
PQ401 98%
PQ401

Caution: for Research Use Only

Formula: C₁₈H₁₆ClN₃O₂
MW: 341.8 g/mol
MDL Number: MFCD00160558
CAS Number: 196868-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR