(±)-Naringenin 98%

Supplier: Ambeed
Warning

Synonyms: (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone, Naringenin

A136202-100G A136202-10G A136202-25G A136202-500G A136202-5G
76984-004EA 92 USD
76984-004 76984-006 76984-008 76984-010 76984-012
(±)-Naringenin 98%
(±)-Naringenin

Caution: for Research Use Only

Formula: C₁₅H₁₂O₅
MW: 272.25 g/mol
MDL Number: MFCD00006844
CAS Number: 67604-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR