Ferrocenecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Ferrocene monocarboxylic acid

A130257-100G A130257-10G A130257-1G A130257-25G A130257-500G A130257-5G
76996-340EA 75 USD
76996-340 76996-342 76996-344 76996-346 76996-348 76996-350
Ferrocenecarboxylic acid 97%
Ferrocenecarboxylic acid

Caution: for Research Use Only

Formula: C₁₁H₁₀FeO₂
MDL Number: MFCD00001430
CAS Number: 1271-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR