Ethyl-4-methoxycinnamate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Methoxycinnamic Acid Ethyl Ester, (E)-Ethyl p-methoxycinnamate, (E)-Ethyl 3-(4-methoxyphenyl)acrylate, Ethyl 3-(4-Methoxyphenyl)acrylate, 3-(4-Methoxyphenyl)acrylic Acid Ethyl Ester
A128415-1G
A128415-250MG
A128415-100G
A128415-25G
A128415-5G
76996-206EA
6
USD
76996-206
76996-208
77599-866
77670-820
77670-822
Ethyl-4-methoxycinnamate 97%
Ethyl-4-methoxycinnamate
Caution: for Research Use Only
Formula:
C₁₂H₁₄O₃ MW: 206.24 g/mol |
MDL Number:
MFCD00026906 CAS Number: 24393-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...