(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxy-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Asp-OMe

A122594-1G A122594-250MG A122594-25G A122594-10G A122594-5G A122594-100G
76995-922EA 8.49 USD
76995-922 76995-924 77571-372 77599-766 77599-768 77670-738
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxy-4-oxobutanoic acid 98%
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxy-4-oxobutanoic acid

Caution: for Research Use Only

Formula: C₂₀H₁₉NO₆
MW: 369.37 g/mol
MDL Number: MFCD03701473
CAS Number: 145038-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR