Sudan IV BS

Supplier: Ambeed
Warning

A120599-100G A120599-500G
76983-228EA 27 USD
76983-228 77611-022
Sudan IV BS
Sudan IV

Caution: for Research Use Only

Formula: C₂₄H₂₀N₄O
MDL Number: MFCD00003893
CAS Number: 85-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR