N-(2-Hydroxyethyl)oleamide 90%
Supplier: Ambeed
|
Warning
|
A115383-1G
A115383-250MG
A115383-25G
A115383-5G
76995-488EA
37
USD
76995-488
76995-490
77670-616
77670-618
N-(2-Hydroxyethyl)oleamide 90%
N-(2-Hydroxyethyl)oleamide
Caution: for Research Use Only
Formula:
C₂₀H₃₉NO₂ |
MDL Number:
MFCD00045972 CAS Number: 111-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...