a-Tosyl(3-methoxybenzyl)isocyanide 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
a-Tosyl-(3-methoxybenzyl) isocyanide
A110618-100MG
A110618-250MG
77025-750EA
45
USD
77025-750
77025-752
a-Tosyl(3-methoxybenzyl)isocyanide 95%
a-Tosyl(3-methoxybenzyl)isocyanide
Caution: for Research Use Only
Formula:
C₁₆H₁₅NO₃S |
MDL Number:
MFCD04114786 CAS Number: 394655-17-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...