α,α,α-Trifluoro-m-toluoyl chloride 96%

Supplier: Ambeed
Danger

Synonyms: 3-(Trifluoromethyl)benzene-1-carbonyl chloride, 3-(Trifluoromethyl)benzoyl chloride

A100725-25G A100725-5G
76994-468EA 16 USD
76994-468 77570-068
α,α,α-Trifluoro-m-toluoyl chloride 96%
α,α,α-Trifluoro-m-toluoyl chloride

Caution: for Research Use Only

Formula: C₈H₄ClF₃O
MW: 208.57
Density: 1.4078
MDL Number: MFCD00000680
CAS Number: 2251-65-2
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR