2,6-Dichloropyridine-4-methanol 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine

A185199-1G A185199-250MG A185199-5G
76974-576EA 6.37 USD
76974-576 76974-578 76974-580 76974-582
2,6-Dichloropyridine-4-methanol 97%
2,6-Dichloropyridine-4-methanol

Caution: for Research Use Only

Formula: C₆H₅Cl₂NO
MDL Number: MFCD00052638
CAS Number: 101990-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR