2,3,5,6-Tetrafluorophenylboronic acid 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2,3,5,6-Tetrafluorophenyl)metaboric acid, (2,3,5,6-Tetrafluorophenyl)boronic acid, Dihydroxy(2,3,5,6-tetrafluorophenyl)borane, 2,3,5,6-Tetrafluorobenzeneboronic acid
A265038-1G
A265038-100MG
A265038-250MG
76977-068EA
146
USD
76977-068
77881-723
77881-724
2,3,5,6-Tetrafluorophenylboronic acid 96%
2,3,5,6-Tetrafluorophenylboronic acid
Caution: for Research Use Only
Formula:
C₆H₃BF₄O₂ MW: 193.89 g/mol |
MDL Number:
MFCD04039310 CAS Number: 511295-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...