Nitrendipine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate

A122241-1G A122241-250MG A122241-50MG A122241-5G A122241-100G A122241-25G A122241-10MG
76971-604EA 25.47 USD
76971-604 76971-606 76971-608 76971-610 77599-756 77599-758 77670-728
Nitrendipine 98%
Nitrendipine

Caution: for Research Use Only

Formula: C₁₈H₂₀N₂O₆
MW: 360.366 g/mol
MDL Number: MFCD00082255
CAS Number: 39562-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR