Dihydrocapsaicin 98%

Supplier: Ambeed
Danger

A862042-100MG A862042-1G A862042-250MG A862042-50MG
76981-834EA 204 USD
76981-834 76981-836 76981-838 76981-840
Dihydrocapsaicin 98%
Dihydrocapsaicin

Caution: for Research Use Only

Formula: C₁₈H₂₉NO₃
MDL Number: MFCD00057794
CAS Number: 19408-84-5
UN: 3462
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR