Gnetol 95%

Supplier: Ambeed
Warning

Synonyms: (E)-2-(3,5-Dihydroxystyryl)benzene-1,3-diol, 2,3',5',6-Tetrahydroxy-trans-stilbene

A449058-10MG A449058-25MG
76979-482EA 90.18 USD
76979-482 76979-484
Gnetol 95%
Gnetol

Caution: for Research Use Only

Formula: C₁₄H₁₂O₄
MW: 244.25 g/mol
MDL Number: MFCD12405548
CAS Number: 86361-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR