Benzo[b]thiophene-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Benzothiophene-3-carboxylic acid

54358-25G 54358-5G
76943-160EA 212.18 USD
76943-160 76943-162
Benzo[b]thiophene-3-carboxylic acid ≥97%
Benzo[b]thiophene-3-carboxylic acid

Caution: for Research Use Only

Formula: C₉H₆O₂S
Storage Temperature: Ambient
MDL Number: MFCD01846406
CAS Number: 5381-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR