Methyl-6-formylpicolinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 6-formylpyridine-2-carboxylate, Methyl 6-formyl-2-pyridinecarboxylate

21280-25G 21280-5G
76937-826EA 275.84 USD
76937-826 76937-828
Methyl-6-formylpicolinate ≥97%
Methyl 6-formylpicolinate

Caution: for Research Use Only

Formula: C₈H₇NO₃
MW: 165.1500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07367925
CAS Number: 69950-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR