2-Methoxy-3-methylpyridine-5-boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
17722-1G
17722-25G
17722-5G
76896-960EA
106.09
USD
76896-960
76896-962
76896-964
2-Methoxy-3-methylpyridine-5-boronic acid ≥97%
2-Methoxy-3-methylpyridine-5-boronic acid
Formula:
C₇H₁₀BNO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD11856046 CAS Number: 1083168-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...