3-Methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

65254-1G 65254-500MG 65254-5G 65254-25G
76917-756EA 527.27 USD
76917-756 76917-758 76917-760 77656-776
3-Methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3-Methyl-[1,1'-biphenyl]-4-carbaldehyde

Caution: for Research Use Only

Formula: C₁₄H₁₂O
MW: 196.2444 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24677709
CAS Number: 1071024-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR