3-(tert-Butyl)-4-ethoxybenzaldehyde ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
63669-1G
63669-25G
63669-500MG
63669-5G
76917-686EA
143.23
USD
76917-686
76917-688
76917-690
76917-692
3-(tert-Butyl)-4-ethoxybenzaldehyde ≥97% research grade
3-(tert-Butyl)-4-ethoxybenzaldehyde
Formula:
C₁₃H₁₈O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD02380390 CAS Number: 681443-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...