3-Chloro-2-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

30400-25G 30400-5G
76913-538EA 275.84 USD
76913-538 76913-540
3-Chloro-2-fluorobenzyl alcohol ≥97%
3-Chloro-2-fluorobenzyl alcohol

Caution: for Research Use Only

Formula: C₇H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD01631549
CAS Number: 261723-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR