1-Bromo-2-(methoxymethoxy)-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

20830-1G 20830-25G 20830-500MG 20830-5G
76884-656EA 168.69 USD
76884-656 76884-658 76884-660 76884-662
1-Bromo-2-(methoxymethoxy)-4-(trifluoromethyl)benzene ≥97%
1-Bromo-2-(methoxymethoxy)-4-(trifluoromethyl)benzene
Formula: C₉H₈BrF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD32706992
CAS Number: 2150186-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR