5-Acetylthiophene-2-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Acetyl-5-formylthiophene, 5-Acetyl-2-thenaldehyde, 5-Acetylthiophene-2-carbaldehyde, 2-Ethanoyl-5-formylthiophene

55050-1G 55050-25G 55050-500MG 55050-5G
76933-526EA 147.47 USD
76933-526 76933-528 76933-530 76933-532
5-Acetylthiophene-2-carboxaldehyde ≥97%
5-Acetylthiophene-2-carboxaldehyde

Caution: for Research Use Only

Formula: C₇H₆O₂S
MW: 154.186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00509280
CAS Number: 4565-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR