4-Formyl-2-methylbenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

14115-1G 14115-25G 14115-500MG 14115-5G
76925-068EA 124.13 USD
76925-068 76925-070 76925-072 76925-074
4-Formyl-2-methylbenzoic acid ≥97% research grade
4-Formyl-2-methylbenzoic acid
Formula: C₉H₈O₃
Storage Temperature: Refrigerator
MDL Number: MFCD20646141
CAS Number: 503470-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR