4-Hexylphenylboronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
12105-25G
12105-5G
76924-676EA
357.53
USD
76924-676
76924-678
4-Hexylphenylboronic acid ≥95%
4-Hexylphenylboronic acid
Caution: for Research Use Only
Formula:
C₁₂H₁₉BO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04039029 CAS Number: 105365-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...