5-Bromo-N,N-diethylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromopyridine-2-carboxylic acid diethylamide

10144-25G 10144-5G
76924-216EA 524.09 USD
76924-216 76924-218
5-Bromo-N,N-diethylpicolinamide ≥97%
5-Bromo-N,N-diethylpicolinamide
Formula: C₁₀H₁₃BrN₂O
MW: 257.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07357420
CAS Number: 673485-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR