(2-Formyl-5-(trifluoromethoxy)phenyl)boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
14110-1G
14110-25G
14110-5G
76895-248EA
70.02
USD
76895-248
76895-250
76895-252
(2-Formyl-5-(trifluoromethoxy)phenyl)boronic acid ≥97%
(2-Formyl-5-(trifluoromethoxy)phenyl)boronic acid
Formula:
C₈H₆BF₃O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD10566600 CAS Number: 1218790-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...