3,4-Dibromo-6-fluoroaniline ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4,5-Dibromo-2-fluoroaniline
69469-1G
69469-25G
69469-5G
76917-880EA
71.09
USD
76917-880
76917-882
76917-884
3,4-Dibromo-6-fluoroaniline ≥97%
3,4-Dibromo-6-fluoroaniline
Formula:
C₆H₄Br₂FN Storage Temperature: Ambient |
MDL Number:
MFCD09878129 CAS Number: 1000578-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...