3-Fluoro-4-hydroxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

68493-1G 68493-500MG 68493-5G 68493-25G
76917-856EA 701.26 USD
76917-856 76917-858 76917-860 77656-812
3-Fluoro-4-hydroxy-2-methylbenzaldehyde ≥97%
3-Fluoro-4-hydroxy-2-methylbenzaldehyde

Caution: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.138 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34763424
CAS Number: 2918889-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR