3-Chloro-2,6-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

16423-25G 16423-5G
76908-488EA 265.23 USD
76908-488 76908-490
3-Chloro-2,6-difluorophenol ≥97%
3-Chloro-2,6-difluorophenol

Caution: for Research Use Only

Formula: C₆H₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD01631330
CAS Number: 261762-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR