HyperSep™ SCX Strong Cation Exchanger SPE Plates
Supplier: Thermo Scientific
60300-582 60300-588
10047-766EA 524.35 USD
10047-766 10047-778
HyperSep™ SCX Strong Cation Exchanger SPE Plates
Microplates Solid Phase Extraction Plates (SPE)
A strong cation sorbent for extraction of charged basic compounds.
  • Particle size: 40 to 63 µm
  • Pore size: 60 Å
  • Surface area: 525 m²/g