HyperSep™ Silica SPE Plates
Supplier: Thermo Scientific
60300-482 60300-485 60300-486 60300-488
10048-408EA 471.89 USD
10048-408 10048-414 10048-416 10048-420
HyperSep™ Silica SPE Plates
Microplates Solid Phase Extraction Plates (SPE)
A polar sorbent primarily used to retain analytes in non-polar matrices.
  • Particle size: 40 to 63 µm
  • Pore size: 60 Å
  • Surface area: 530 m²/g